(2,4-Bis(trifluoromethyl)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Bis(trifluoromethyl)phenylacetic acid

A229204-100MG A229204-250MG A229204-5G
76771-084EA 6.37 USD
76771-084 76771-086 76771-088
(2,4-Bis(trifluoromethyl)phenyl)acetic acid 97%
(2,4-Bis(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(CF₃)₂)OCH₂COOH
MW: 272.15 g/mol
MDL Number: MFCD00042492
CAS Number: 177952-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR