6-methoxyquinoxaline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxyquinoxaline

A228790-100MG A228790-1G A228790-250MG A228790-5G
76771-076EA 20.16 USD
76771-076 76771-078 76771-080 76771-082
6-methoxyquinoxaline 98%
6-methoxyquinoxaline

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.176 g/mol
MDL Number: MFCD00666601
CAS Number: 6639-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR