(Chloromethyl)diethoxy(methyl)silane 97%

Supplier: Ambeed
Danger

Synonyms: Chloromethylmethyldiethoxysilane

A228004-5G A228004-25G
76771-048EA 23 USD
76771-048 77626-176
(Chloromethyl)diethoxy(methyl)silane 97%
(Chloromethyl)diethoxy(methyl)silane

Precaución: for Research Use Only

Formula: C₆H₁₅ClO₂Si
MW: 182.72 g/mol
MDL Number: MFCD00018878
CAS Number: 2212-10-4
UN: 2924
ADR: 3,8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR