4-Bromo-2-chlorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A226488-100G
A226488-10G
A226488-1G
A226488-5G
A226488-25G
76770-998EA
90.18
USD
76770-998
76771-000
76771-002
76771-004
77626-166
4-Bromo-2-chlorobenzoic acid 98%
4-Bromo-2-chlorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol |
MDL Number:
MFCD00040903 CAS Number: 59748-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...