6-Morpholino-3-pyridinamine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine

A224552-1G A224552-5G
76770-970EA 79 USD
76770-970 76770-972
6-Morpholino-3-pyridinamine 97%
6-Morpholino-3-pyridinamine

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O
MW: 179.22 g/mol
MDL Number: MFCD00119562
CAS Number: 52023-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR