3-Amino-6-chloropyridine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-6-chloropicolinamide

A224341-100MG A224341-10G A224341-1G A224341-250MG A224341-25G A224341-5G
76770-958EA 26.53 USD
76770-958 76770-960 76770-962 76770-964 76770-966 76770-968
3-Amino-6-chloropyridine-2-carboxamide 98%
3-Amino-6-chloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD09750671
CAS Number: 175358-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR