3-Phenylpropyl carbamate 97%

Supplier: Ambeed
Warning

Synonyms: Phenprobamate

A224166-1G A224166-250MG A224166-5G A224166-25G
76770-944EA 9 USD
76770-944 76770-946 76770-948 77877-696
3-Phenylpropyl carbamate 97%
3-Phenylpropyl carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00868163
CAS Number: 673-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR