2-Fluoro-4-nitrotoluene 98%
Supplier: Ambeed
|
Warning
|
A224064-100G
A224064-10G
A224064-25G
A224064-500G
A224064-5G
76770-934EA
72.15
USD
76770-934
76770-936
76770-938
76770-940
76770-942
2-Fluoro-4-nitrotoluene 98%
2-Fluoro-4-nitrotoluene
Precaución: for Research Use Only
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol |
MDL Number:
MFCD00007199 CAS Number: 1427-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...