5-Bromo-6-phenyl-3(2H)-pyridazinone 97%

Supplier: Ambeed
Warning

A223958-10G A223958-1G A223958-25G A223958-5G
76770-926EA 253 USD
76770-926 76770-928 76770-930 76770-932
5-Bromo-6-phenyl-3(2H)-pyridazinone 97%
5-Bromo-6-phenyl-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂O
MDL Number: MFCD02083298
CAS Number: 90766-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR