5-Iodo-1H-benzimidazole 97%
Supplier: Ambeed
|
Warning
|
A223044-100MG
A223044-1G
A223044-250MG
A223044-5G
76770-912EA
25
USD
76770-912
76770-914
76770-916
76770-918
5-Iodo-1H-benzimidazole 97%
5-Iodo-1H-benzimidazole
Precaución: for Research Use Only
Formula:
C₇H₅IN₂ |
MDL Number:
MFCD06659631 CAS Number: 78597-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...