Ethyl-4,4,4-trifluorobutyrate 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate

A222990-1G A222990-5G
76770-902EA 14.86 USD
76770-902 76770-904
Ethyl-4,4,4-trifluorobutyrate 98%
Ethyl-4,4,4-trifluorobutyrate

Precaución: for Research Use Only

Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
MDL Number: MFCD00041398
CAS Number: 371-26-6
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR