5-Oxo-5-(pyrrolidin-1-yl)pentanoic acid 95%

Supplier: Ambeed
Warning

A222894-1G A222894-250MG A222894-5G
76770-896EA 37 USD
76770-896 76770-898 76770-900
5-Oxo-5-(pyrrolidin-1-yl)pentanoic acid 95%
5-Oxo-5-(pyrrolidin-1-yl)pentanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MDL Number: MFCD01339572
CAS Number: 5724-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR