Methyl-5-bromothiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-thiophene-2-carboxylic acid methyl ester, Methyl 5-bromothiophene-2-carboxylate

A222575-10G A222575-1G A222575-25G A222575-5G
76770-886EA 333.13 USD
76770-886 76770-888 76770-890 76770-892 76770-894
Methyl-5-bromothiophene-2-carboxylate 98%
Methyl-5-bromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S
MDL Number: MFCD03233845
CAS Number: 62224-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR