5-Chloro-2-hydroxy-3-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-methylpyridin-2-ol

A222565-10G A222565-1G A222565-250MG A222565-5G
76770-878EA 195.21 USD
76770-878 76770-880 76770-882 76770-884
5-Chloro-2-hydroxy-3-methylpyridine 95%
5-Chloro-2-hydroxy-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD00126916
CAS Number: 58498-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR