6,7-Dichloroquinoxaline-2,3-dione 97%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dichloroquinoxaline-2,3(1H,4H)-dione, 6,7-Dichloro-1,4-dihydroquinoxaline-2,3-dione

A222181-1G A222181-25G A222181-5G
76770-862EA 16.98 USD
76770-862 76770-864 76770-866
6,7-Dichloroquinoxaline-2,3-dione 97%
6,7-Dichloroquinoxaline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂O₂
MW: 231.0356 g/mol
MDL Number: MFCD00078575
CAS Number: 25983-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR