1,3-Diiodo-5,5-dimethylhydantoin 97%

Supplier: Ambeed
Danger

A221815-25G A221815-5G
76770-852EA 173 USD
76770-852 76770-854
1,3-Diiodo-5,5-dimethylhydantoin 97%
1,3-Diiodo-5,5-dimethylhydantoin

Precaución: for Research Use Only

Formula: C₅H₆I₂N₂O₂
MDL Number: MFCD00020867
CAS Number: 2232-12-4
UN: 3085
ADR: 5.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR