5-Bromo-1-methyl-1H-benzo[d]Imidazole 95%

Supplier: Ambeed
Warning

A221529-100MG A221529-10G A221529-1G A221529-250MG A221529-25G A221529-5G
76770-836EA 18.04 USD
76770-836 76770-838 76770-840 76770-842 76770-844 76770-846
5-Bromo-1-methyl-1H-benzo[d]Imidazole 95%
5-Bromo-1-methyl-1H-benzo[d]Imidazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.05 g/mol
MDL Number: MFCD09878409
CAS Number: 53484-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR