3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-(((tert-Butoxycarbonyl)amino)methyl)phenyl)boronic acid, 3-(Aminomethyl)benzeneboronic acid, N-BOC protected

A221510-100MG A221510-1G A221510-250MG A221510-5G
76770-828EA 27.59 USD
76770-828 76770-830 76770-832 76770-834
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄
MW: 251.09 g/mol
MDL Number: MFCD06246052
CAS Number: 199609-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR