α,α,α-Trifluoro-m-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)benzoic acid

A221256-1000G A221256-100G A221256-10G A221256-25G A221256-500G
76770-806EA 149 USD
76770-806 76770-808 76770-810 76770-812 76770-814 76770-816
α,α,α-Trifluoro-m-toluic acid 98%
α,α,α-Trifluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MDL Number: MFCD00002519
CAS Number: 454-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR