5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-tert-butyl-1h-pyrazole-4-carbonitrile, 5-Amino-1-(tert-butyl)-1H-pyrazole-4-carbonitrile

A220569-100MG A220569-1G A220569-250MG A220569-5G
76770-794EA 25.47 USD
76770-794 76770-796 76770-798 76770-800
5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₁₂N₄
MW: 164.21 g/mol
MDL Number: MFCD00128289
CAS Number: 158001-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR