(5-Fluoropyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-(hydroxymethyl)pyridine

A219778-100MG A219778-10G A219778-1G A219778-250MG A219778-5G
76770-772EA 20.16 USD
76770-772 76770-774 76770-776 76770-778 76770-780
(5-Fluoropyridin-2-yl)methanol 97%
(5-Fluoropyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.1200 g/mol
MDL Number: MFCD09027049
CAS Number: 802325-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR