5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine 95%

Supplier: Ambeed
Warning

A219722-10G A219722-1G A219722-250MG A219722-25G
76770-758EA 88 USD
76770-758 76770-760 76770-762 76770-764
5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine 95%
5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃S
MDL Number: MFCD02032690
CAS Number: 100987-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR