Ethyl 3-(benzo[d][1,3]dioxol-5-yl)-3-oxopropanoate 98%

Supplier: Ambeed
Warning

A219639-100MG A219639-1G
76770-754EA 81.69 USD
76770-754 76770-756
Ethyl 3-(benzo[d][1,3]dioxol-5-yl)-3-oxopropanoate 98%
Ethyl 3-(benzo[d][1,3]dioxol-5-yl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₅
MW: 236.22 g/mol
MDL Number: MFCD01463468
CAS Number: 81581-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR