2,5-Dimethoxyphenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2,5-Dimethoxyphenyl)metaboric acid, 2,5-Dimethoxybenzeneboronic acid, (2,5-Dimethoxyphenyl)boronic acid, Dihydroxy(2,5-dimethoxyphenyl)borane
A217546-100G
A217546-10G
A217546-1G
A217546-25G
A217546-5G
76770-736EA
169.75
USD
76770-736
76770-738
76770-740
76770-742
76770-744
2,5-Dimethoxyphenylboronic acid 97%
2,5-Dimethoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₁BO₄ MW: 181.98 g/mol |
MDL Number:
MFCD01318181 CAS Number: 107099-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...