5-Methyl-1H-pyrrole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A216724-250MG A216724-1G A216724-5G
76770-708EA 36 USD
76770-708 77626-092 77877-678
5-Methyl-1H-pyrrole-2-carboxylic acid 98%
5-Methyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD03011381
CAS Number: 3757-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR