6-Bromo-3-methyl-1H-indazole 98%

Supplier: Ambeed
Warning

A216434-1G A216434-250MG A216434-5G A216434-10G A216434-25G
76770-692EA 35.01 USD
76770-692 76770-694 76770-696 77626-088 77626-090
6-Bromo-3-methyl-1H-indazole 98%
6-Bromo-3-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD08272244
CAS Number: 7746-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR