5-Bromo-4-chloro-2-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A216381-1G A216381-250MG A216381-25G A216381-5G
76770-684EA 28.65 USD
76770-684 76770-686 76770-688 76770-690
5-Bromo-4-chloro-2-fluorobenzoic acid 97%
5-Bromo-4-chloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
MDL Number: MFCD00672928
CAS Number: 289038-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR