4-Bromo-α,α,α,2-tetrafluorotoluene 98%

Supplier: Ambeed
Warning

A215906-100G A215906-10G A215906-1G A215906-25G A215906-5G
76770-672EA 130 USD
76770-672 76770-674 76770-676 76770-678 76770-680
4-Bromo-α,α,α,2-tetrafluorotoluene 98%
4-Bromo-α,α,α,2-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MDL Number: MFCD01075254
CAS Number: 142808-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR