7-Methoxy-2-tetralone 98%

Supplier: Ambeed
Warning

A215873-100G A215873-1G A215873-250MG A215873-25G A215873-10G A215873-5G
76770-666EA 251 USD
76770-666 76770-668 76770-670 77593-408 77626-086 77877-676
7-Methoxy-2-tetralone 98%
7-Methoxy-2-tetralone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MDL Number: MFCD00001730
CAS Number: 4133-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR