2,4-Dimethyl-5-thiazolecarboxylic acid 98%

Supplier: Ambeed
Warning

A214902-10G A214902-1G A214902-25G A214902-5G
76770-648EA 44 USD
76770-648 76770-650 76770-652 76770-654
2,4-Dimethyl-5-thiazolecarboxylic acid 98%
2,4-Dimethyl-5-thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MDL Number: MFCD00052943
CAS Number: 53137-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR