5H-Dibenzo[b,e]azepine-6,11-dione 98%

Supplier: Ambeed
Warning

A213639-5G A213639-1G
76770-628EA 498.63 USD
76770-628 77600-722
5H-Dibenzo[b,e]azepine-6,11-dione 98%
5H-Dibenzo[b,e]azepine-6,11-dione

Precaución: for Research Use Only

Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
MDL Number: MFCD00612476
CAS Number: 1143-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR