2-Amyl-5-methylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-pentylphenol, Amylmetacresol

A213022-1G A213022-5G
76770-606EA 16.98 USD
76770-606 76770-608
2-Amyl-5-methylphenol 98%
2-Amyl-5-methylphenol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
MW: 178.28 g/mol
MDL Number: MFCD00127710
CAS Number: 1300-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR