3-Chloro-6-(ethylamino)pyridazine 97%

Supplier: Ambeed
Danger

A212952-1G A212952-5G
76770-602EA 12 USD
76770-602 76770-604
3-Chloro-6-(ethylamino)pyridazine 97%
3-Chloro-6-(ethylamino)pyridazine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MDL Number: MFCD08363097
CAS Number: 68588-39-6
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR