2,4-Dinitrobenzaldehyde 98%

Supplier: Ambeed
Danger

A212395-25G A212395-5G
76770-582EA 27.59 USD
76770-582 76770-584
2,4-Dinitrobenzaldehyde 98%
2,4-Dinitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄N₂O₅
MW: 196.12 g/mol
MDL Number: MFCD00013376
CAS Number: 528-75-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR