Glibenclamide 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulphamoyl)phenethyl)-2-methoxybenzamide, 5-Chloro-N-[4-(3-cyclohexylureidosulphonyl)phenethyl]-2-methoxybenzamide, Glyburide

A212350-1G A212350-1MG A212350-25G A212350-5G A212350-100G
76770-574EA 6.37 USD
76770-574 76770-576 76770-578 76770-580 77598-322
Glibenclamide 98%
Glibenclamide

Precaución: for Research Use Only

Formula: C₂₃H₂₈ClN₃O₅S
MW: 494.00 g/mol
MDL Number: MFCD00056625
CAS Number: 10238-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR