Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Amino-4-thiazolyl)-2-oxoacetic acid ethyl ester, Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate, Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate

A212347-1G A212347-500G A212347-25G A212347-100G
76770-570EA 6 USD
76770-570 76770-572 77568-048 77877-665
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate 97%
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate

Precaución: For research use only.

Formula: C₇H₉NO₄S
MDL Number: MFCD00010414
CAS Number: 64987-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR