5-Bromo-2-(methylthio)pyridine 98%

Supplier: Ambeed
Danger

A212307-100G A212307-10G A212307-25G A212307-5G A212307-1G
76770-562EA 280 USD
76770-562 76770-564 76770-566 76770-568 77626-074
5-Bromo-2-(methylthio)pyridine 98%
5-Bromo-2-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNS
MDL Number: MFCD03411571
CAS Number: 51933-78-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR