2-Fluoro-5-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

A211627-10G A211627-1G A211627-25G A211627-5G
76770-544EA 39.26 USD
76770-544 76770-546 76770-548 76770-550
2-Fluoro-5-methylphenylboronic acid 97%
2-Fluoro-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂
MW: 153.9466 g/mol
MDL Number: MFCD03428512
CAS Number: 166328-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR