5-Bromo-N,N-dimethylnicotinamide 95%

Supplier: Ambeed
Warning

A211231-10G A211231-1G A211231-5G
76770-518EA 187 USD
76770-518 76770-520 76770-522
5-Bromo-N,N-dimethylnicotinamide 95%
5-Bromo-N,N-dimethylnicotinamide

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O
MDL Number: MFCD01788370
CAS Number: 292170-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR