5-Bromo-2-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A210816-1G A210816-25G A210816-5G
76770-504EA 9 USD
76770-504 76770-506 77877-660
5-Bromo-2-iodobenzonitrile 95%
5-Bromo-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
MDL Number: MFCD00672950
CAS Number: 121554-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR