2-Amino-5-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

A210157-100G A210157-10G A210157-1G A210157-250MG A210157-25G A210157-5G
76770-474EA 263 USD
76770-474 76770-476 76770-478 76770-480 76770-482 76770-484
2-Amino-5-fluorobenzonitrile 97%
2-Amino-5-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MDL Number: MFCD02683869
CAS Number: 61272-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR