3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide 97%

Supplier: Ambeed
Warning

Synonyms: 3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide, Thiazolium, 3-ethyl-5-(2-hydroxyethyl)-4-methyl-, bromide

A210112-100G A210112-10G A210112-25G A210112-500G A210112-5G
76770-464EA 133.68 USD
76770-464 76770-466 76770-468 76770-470 76770-472
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide 97%
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide

Precaución: for Research Use Only

Formula: C₈H₁₄BrNOS
MW: 252.16 g/mol
MDL Number: MFCD00040549
CAS Number: 54016-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR