Itaconic anhydride 99%

Supplier: Ambeed
Warning

Synonyms: Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione

A209795-100G A209795-500G
76770-454EA 13 USD
76770-454 77598-318
Itaconic anhydride 99%
Itaconic anhydride

Precaución: for Research Use Only

Formula: C₅H₄O₃
MDL Number: MFCD00005530
CAS Number: 2170-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR