4-(4-Chlorophenoxy)aniline 97%

Supplier: Ambeed
Danger

A209725-1G A209725-5G A209725-10G A209725-25G
76770-450EA 9 USD
76770-450 76770-452 77568-022 77877-658
4-(4-Chlorophenoxy)aniline 97%
4-(4-Chlorophenoxy)aniline

Precaución: For research use only.

Formula: C₁₂H₁₀ClNO
MDL Number: MFCD00043925
CAS Number: 101-79-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR