5-(4-Nitrophenyl)-1H-pyrazole-3-carboxylic acid 97+%

Supplier: Ambeed
Warning

A209178-1G A209178-250MG A209178-5G
76770-430EA 66.84 USD
76770-430 76770-432 76770-434
5-(4-Nitrophenyl)-1H-pyrazole-3-carboxylic acid 97+%
5-(4-Nitrophenyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇N₃O₄
MW: 233.1803 g/mol
MDL Number: MFCD03086935
CAS Number: 189083-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR