α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

A209020-100G A209020-10G A209020-500G A209020-5G A209020-25G
76770-416EA 26.53 USD
76770-416 76770-418 76770-420 76770-422 77086-932
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine

Precaución: for Research Use Only

Formula: C₈H₅F₆N
MW: 229.12 g/mol
MDL Number: MFCD00000394
CAS Number: 328-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR