2-Amino-5-bromo-4,6-dimethylpyrimidine 97%

Supplier: Ambeed
Warning

A208713-10G A208713-1G A208713-250MG A208713-25G
76770-402EA 103 USD
76770-402 76770-404 76770-406 77877-654
2-Amino-5-bromo-4,6-dimethylpyrimidine 97%
2-Amino-5-bromo-4,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₈BrN₃
MDL Number: MFCD00160498
CAS Number: 4214-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR