5-Methyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A208623-100MG A208623-1G A208623-250MG A208623-25G A208623-5G
76770-392EA 22 USD
76770-392 76770-394 76770-396 76770-398 76770-400
5-Methyl-1H-pyrrolo[2,3-b]pyridine 95%
5-Methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C8H8N2
MDL Number: MFCD06659674
CAS Number: 824-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR