3-Bromo-2,4,5-trifluorobenzoic acid 98%

Supplier: Ambeed
Warning

A208308-1G A208308-5G
76770-388EA 39.26 USD
76770-388 76770-390
3-Bromo-2,4,5-trifluorobenzoic acid 98%
3-Bromo-2,4,5-trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂BrF₃O₂
MW: 254.99 g/mol
MDL Number: MFCD05664065
CAS Number: 104222-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR