2-Phenoxyaniline 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-2-phenoxybenzol

A207779-100G A207779-10G A207779-25G A207779-500G
76770-368EA 44.56 USD
76770-368 76770-370 76770-372 76770-374
2-Phenoxyaniline 98%
2-Phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00035765
CAS Number: 2688-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR